2-Bromostyrene ≥97% stabilized

Supplier: BeanTown Chemical

217610-25G 217610-5G
BT217610-25GEA 558 USD
BT217610-25G BT217610-5G
2-Bromostyrene ≥97% stabilized
2-Bromostyrene

CAS: 2039-88-5; EC No: 218-027-3; MDL No: MFCD00000076 Liquid; Molecular Formula: C8H7Br; MW: 183.05 Boiling Point: 102-104°/22 mmHg; Flash point: 86°C (187°F) Density (g/mL): 1.46; Refractive Index: 1.592


  • Refractive index: 1.592
  • Liquid

Formula: C₈H₇Br
MW: 183.05 g/mol
Boiling Pt: 102…104 °C/22 mmHg
Density: 1.46 g/mL
Flash Pt: 86 °C (187 °F)
MDL Number: MFCD00000076
CAS Number: 2039-88-5
EINECS: 218-027-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR