Perfluoro(methylcyclohexane) ≥94%

Supplier: BeanTown Chemical

Synonyms: FlutecR PP2

217600-100G 217600-25G 217600-500G
BT217600-100GEA 219 USD
BT217600-100G BT217600-25G BT217600-500G
Perfluoro(methylcyclohexane) ≥94%
Perfluoro(methylcyclohexane)

CAS: 355-02-2; EC No: 206-573-5; MDL No: MFCD00001468; RTECS: GV7610000 Liquid; Linear Formula: C6F11CF3; MW: 350.06 Melting Point: -70°; Boiling Point: 75-76° Density (g/mL): 1.787; Refractive Index: 1.285


  • RTECS: GV7610000
  • Refractive index: 1.285
  • Liquid

Formula: C₇F₁₄
MW: 350.05 g/mol
Boiling Pt: 75…76 °C
Melting Pt: –70 °C
Density: 1.787 g/mL
MDL Number: MFCD00001468
CAS Number: 355-02-2
EINECS: 206-573-5

Order Now

Specification Test Results

Assay 94%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR