EDTA (ethylenediamine tetraacetic acid) ≥99%
Supplier: BeanTown Chemical
Synonyms:
Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
217585-1KG
217585-500G
217585-5KG
BT217585-1KGEA
63
USD
BT217585-1KG
BT217585-500G
BT217585-5KG
EDTA (ethylenediamine tetraacetic acid) ≥99%
EDTA (ethylenediamine tetraacetic acid)
CAS: 60-00-4; EC No: 200-449-4; MDL No: MFCD00003541; RTECS: AH4025000
Powder; Molecular Formula: C10H16N2O8; MW: 292.24
Melting Point: 250° (decomposes)
- RTECS: AH4025000
- Powder
Formula:
C₁₀H₁₆N₂O₈ MW: 292.25 g/mol Melting Pt: 250 °C (dec.) |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 EINECS: 200-449-4 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...