Ethyl-3-phenylpropionate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethyl 3-Phenylpropionate, Ethyl hydrocinnamate, 3-Phenylpropionic acid ethyl ester, Ethyl dihydrocinnamate, Hydrocinnamic acid ethyl ester

217525-1KG 217525-250G 217525-50G
BT217525-1KGEA 920 USD
BT217525-1KG BT217525-250G BT217525-50G
Ethyl-3-phenylpropionate ≥98%
Ethyl-3-phenylpropionate

CAS: 2021-28-5; EC No: 217-966-6; MDL No: MFCD00009206 Liquid; Linear Formula: C6H5CH2CH2CO2CH2CH3; Molecular Formula: C11H14O2; MW: 178.23 Boiling Point: 247-248°; Flash point: 108°C (226°F) Density (g/mL): 1.01; Refractive Index: 1.494


  • Refractive index: 1.494
  • Liquid

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 247…248 °C
Density: 1.01 g/mL
Flash Pt: 108 °C (226 °F)
MDL Number: MFCD00009206
CAS Number: 2021-28-5
EINECS: 217-966-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR