1-Bromononane ≥99%
Supplier: BeanTown Chemical
Synonyms:
1-Nonyl bromide
217495-100G
217495-25G
217495-500G
BT217495-100GEA
105
USD
BT217495-100G
BT217495-25G
BT217495-500G
1-Bromononane ≥99%
1-Bromononane
CAS: 693-58-3; EC No: 211-755-2; MDL No: MFCD00000278
Liquid; Linear Formula: CH3(CH2)8Br; MW: 207.15
Boiling Point: 201°; Flash point: 90°C (194°F)
Density (g/mL): 1.084; Refractive Index: 1.454
- Refractive index: 1.454
- Liquid
Formula:
BrH₂C(CH₂)₇CH₃ Boiling Pt: 201 °C Density: 1.084 g/mL Flash Pt: 90 °C (194 °F) |
MDL Number:
MFCD00000278 CAS Number: 693-58-3 EINECS: 211-755-2 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...