MOPS-Na ≥98%

Supplier: BeanTown Chemical

217480-100G 217480-25G
BT217480-100GEA 110.75 USD
BT217480-100G BT217480-25G
MOPS-Na ≥98%
MOPS-Na

CAS: 71119-22-7; MDL No: MFCD00064350 Powder; Molecular Formula: C7H14NNaO4S ; MW: 231.25 Melting Point: 221° Density (g/mL): 1.41


  • Powder

Formula: C₇H₁₄NNaO₄S
Melting Pt: 221 °C
Density: 1.41 g/mL
MDL Number: MFCD00064350
CAS Number: 71119-22-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR