2-Amino-5-bromopyridine ≥98%

Supplier: BeanTown Chemical

217440-100G 217440-25G 217440-5G
BT217440-100GEA 348 USD
BT217440-100G BT217440-25G BT217440-5G
2-Amino-5-bromopyridine ≥98%
2-Amino-5-bromopyridine

CAS: 1072-97-5; EC No: 214-019-9; MDL No: MFCD00006323 Crystalline; Molecular Formula: C5H5BrN2 ; MW: 173.01 Melting Point: 133-138°


  • Crystalline

Formula: C₅H₅BrN₂
Melting Pt: 133…138 °C
MDL Number: MFCD00006323
CAS Number: 1072-97-5
EINECS: 214-019-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR