Molybdenum(II) acetate dimer

Supplier: BeanTown Chemical

217380-1G 217380-25G 217380-5G
BT217380-1GEA 66 USD
BT217380-1G BT217380-25G BT217380-5G
Molybdenum(II) acetate dimer
Molybdenum(II) acetate dimer

CAS: 14221-06-8; EC No: 238-089-5; MDL No: MFCD00013395 Crystalline; Linear Formula: [Mo(OCOCH3)2]2; Molecular Formula: C4H12Mo2O4; MW: 428.06 Air Sensitive, Light Sensitive


  • Air Sensitive, Light Sensitive
  • Crystalline

Formula: C₈H₁₂Mo₂O₈
MDL Number: MFCD00013395
CAS Number: 14221-06-8
EINECS: 238-089-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR