4-(3-Chloropropyl)morpholine

Supplier: BeanTown Chemical

Synonyms: 3-Morpholinopropyl Chloride

217350-10G 217350-50G
BT217350-10GEA 65 USD
BT217350-10G BT217350-50G
4-(3-Chloropropyl)morpholine
4-(3-Chloropropyl)morpholine

CAS: 7357-67-7; EC No: 461-510-0; MDL No: MFCD00035333; RTECS: QE0488000 Liquid; Molecular Formula: C7H14ClNO; MW: 163.65


  • RTECS: QE0488000
  • Liquid

Formula: C₇H₁₄ClNO
MW: 163.65 g/mol
MDL Number: MFCD00039714
CAS Number: 7357-67-7
EINECS: 461-510-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR