N-Boc-L-prolinal ≥96%
Supplier: BeanTown Chemical
Synonyms:
(S)-1-Boc-2-Formylpyrrolidine
217340-1G
217340-5G
BT217340-1GEA
155
USD
BT217340-1G
BT217340-5G
N-Boc-L-prolinal ≥96%
N-Boc-L-prolinal
CAS: 69610-41-9; MDL No: MFCD00274186
UN No: UN1989; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C10H17NO3; MW: 199.25
Boiling Point: 211°; Flash point: 57°C (135°F)
Density (g/mL): 1.063; Refractive Index: 1.462; Optical Rotation: [α]20/D -101°, c = 0.66 in chloroform
Air Sensitive
- Optical rotation: [α]20/D -101°, c = 0.66 in chloroform
- Refractive index: 1.462
- Air Sensitive
- Liquid
Formula:
C₁₀H₁₇NO₃ MW: 199.25 g/mol Boiling Pt: 211 °C Density: 1.063 g/mL Flash Pt: 57 °C (135 °F) |
MDL Number:
MFCD00274186 CAS Number: 69610-41-9 ADR: 3,III |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...