5-Bromo-2-methoxynicotinic acid ≥97%

Supplier: BeanTown Chemical

217330-1G 217330-5G
BT217330-1GEA 220 USD
BT217330-1G BT217330-5G
5-Bromo-2-methoxynicotinic acid ≥97%
5-Bromo-2-methoxynicotinic acid

CAS: 54916-66-4; MDL No: MFCD06656771 Solid; Molecular Formula: C7H6BrNO3; MW: 232.03


  • Solid

Formula: C₇H₆BrNO₃
MW: 232.04 g/mol
MDL Number: MFCD06656771
CAS Number: 54916-66-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR