Vinylene carbonate ≥97% stabilized

Supplier: BeanTown Chemical

Synonyms: 1,3-Dioxol-2-one

217315-10G 217315-2G 217315-500MG
BT217315-10GEA 260 USD
BT217315-10G BT217315-2G BT217315-500MG
Vinylene carbonate ≥97% stabilized
Vinylene carbonate

CAS: 872-36-6; EC No: 212-825-5; MDL No: MFCD00005380 Liquid; Molecular Formula: C3H2O3; MW: 86.05 Melting Point: 19-22°; Boiling Point: 162°; Flash point: 73°C (163°F) Density (g/mL): 1.355; Refractive Index: 1.421


  • Refractive index: 1.421
  • Liquid

Formula: C₃H₂O₃
MW: 86.05 g/mol
Boiling Pt: 162 °C
Melting Pt: 19…22 °C
Density: 1.355 g/mL
Flash Pt: 73 °C (163 °F)
MDL Number: MFCD00005380
CAS Number: 872-36-6
EINECS: 212-825-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR