Aluminium oxide activated neutral Brockmann Grade I 58 angstroms

Supplier: BeanTown Chemical

217310-1KG 217310-250G 217310-5KG
BT217310-1KGEA 95.67 USD
BT217310-1KG BT217310-250G BT217310-5KG
Aluminium oxide activated neutral Brockmann Grade I 58 angstroms
Aluminium oxide activated neutral

CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424 -60 Mesh Powder, S.A. 150 m2/g; Molecular Formula: Al2O3; MW: 101.96 Melting Point: 2040°


  • -60 Mesh Powder, S.A. 150 m²/g

Formula: Al₂O₃
MW: 101.96 g/mol
Melting Pt: 2040 °C
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR