Gallic acid monohydrate ≥98.0% ACS

Supplier: BeanTown Chemical

Synonyms: 3,4,5-Trihydroxybenzoic acid monohydrate, 3,4,5-Trihydroxybenzoic acid hydrate

217295-100G 217295-500G
BT217295-100GEA 53 USD
BT217295-100G BT217295-500G
Gallic acid monohydrate ≥98.0% ACS
Gallic acid monohydrate

CAS: 5995-86-8; EC No: 205-749-9; MDL No: MFCD00149098 Powder; Molecular Formula: C7H6O5·H2O; MW: 188.13 Melting Point: 252° (decomposes) Light Sensitive


  • Light Sensitive
  • Powder

Formula: C₇H₆O₅·1H₂O
MW: 188.14 g/mol
Melting Pt: 252 °C (dec.)
MDL Number: MFCD00149098
CAS Number: 5995-86-8
EINECS: 205-749-9

Order Now

Specification Test Results

Assay 98.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR