Potassium hydrogen carbonate ≥99%

Supplier: BeanTown Chemical

Synonyms: Potassium bicarbonate

217260-10KG 217260-2.5KG 217260-500G
BT217260-10KGEA 301 USD
BT217260-10KG BT217260-2.5KG BT217260-500G
Potassium hydrogen carbonate ≥99%
Potassium hydrogen carbonate

CAS: 298-14-6; EC No: 206-059-0; MDL No: MFCD00011402 Crystalline; Linear Formula: KHCO3; MW: 100.12 Density (g/mL): 2.17 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: KHCO₃
MW: 100.12 g/mol
Melting Pt: 292
Density: 2.17 g/mL
MDL Number: MFCD00011402
CAS Number: 298-14-6
EINECS: 206-059-0
Merck Index: 13,07691

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR