Titanium ≥99.995% (trace metals basis), slug, Length 6.35 mm, Diameter 3.175 mm
Supplier: BeanTown Chemical
217230-25G
217230-5G
BT217230-25GEA
159
USD
BT217230-25G
BT217230-5G
Titanium ≥99.995% (trace metals basis), slug, Length 6.35 mm, Diameter 3.175 mm
Titanium
CAS: 7440-32-6; EC No: 231-142-3; MDL No: MFCD00011264
Solid; Molecular Formula: Ti; MW: 47.87
Melting Point: 1660°; Boiling Point: 3287°
Density (g/mL): 4.5
- Solid
Formula:
Ti MW: 47.88 g/mol Boiling Pt: 3287 °C Melting Pt: 1660 °C Density: 4.5 g/mL |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 |
Specification Test Results
Assay | 99.995% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...