(R)-(-)-Mandelic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid
217220-100G
217220-25G
217220-5G
BT217220-100GEA
238
USD
BT217220-100G
BT217220-25G
BT217220-5G
(R)-(-)-Mandelic acid ≥98%
(R)-(-)-Mandelic acid
CAS: 611-71-2; EC No: 210-276-6; MDL No: MFCD00064251
Crystalline; Molecular Formula: C8H8O3; MW: 152.15
Melting Point: 131-133°
Optical Rotation: [α]25/D −151°, c = 1 in ethanol
Light Sensitive
- Optical rotation: [α]25/D −151°, c = 1 in ethanol
- Light Sensitive
- Crystalline
Formula:
C₈H₈O₃ Melting Pt: 131…133 °C |
MDL Number:
MFCD00064251 CAS Number: 611-71-2 EINECS: 210-276-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...