4-Cyano-3-fluorophenyl-trans-4-(4-n-propylcyclohexyl)benzoate ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Cyano-3-fluorophenyl 4-(trans-4-propylcyclohexyl)benzoate

217195-1G 217195-5G
BT217195-1GEA 103 USD
BT217195-1G BT217195-5G
4-Cyano-3-fluorophenyl-trans-4-(4-n-propylcyclohexyl)benzoate ≥97%
4-Cyano-3-fluorophenyl-trans-4-(4-n-propylcyclohexyl)benzoate

CAS: 92118-82-6; MDL No: MFCD11053365 Powder; Molecular Formula: C23H24FNO2; MW: 365.45 Melting Point: 99-101°


  • Powder

Formula: C₂₃H₂₄FNO₂
MW: 365.448 g/mol
Melting Pt: 99…101 °C
MDL Number: MFCD11053365
CAS Number: 92118-82-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR