Tin ≥99.95% (trace metals basis), wire Ø 3.2 mm
Supplier: BeanTown Chemical
217155-1M
217155-5M
BT217155-1MEA
90
USD
BT217155-1M
BT217155-5M
Tin ≥99.95% (trace metals basis), wire Ø 3.2 mm
Tin
CAS: 7440-31-5; EC No: 231-141-8; MDL No: MFCD00133862
Solid; Molecular Formula: Sn; MW: 118.71
Melting Point: 231.9°; Boiling Point: 2270°
Density (g/mL): 7.31
- Solid
Formula:
Sn MW: 118.71 g/mol Boiling Pt: 2270 Melting Pt: 232 Density: 7.31 g/mL |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 Merck Index: 12,09587 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...