Potassium molybdate(VI) ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Potassium molybdate, Molybdic acid potassium salt, Dipotassium tetraoxomolybdate, Molybdic acid, potassium salt, Potassium molybdenum oxide, di-Potassium molybdate

217150-10G 217150-50G
BT217150-10GEA 44 USD
BT217150-10G BT217150-50G
Potassium molybdate(VI) ≥99.9% (trace metals basis)
Potassium molybdate(VI)

CAS: 13446-49-6; EC No: 236-599-2; MDL No: MFCD00011370 -200 Mesh Powder; Molecular Formula: K2MoO4; MW: 238.14 Melting Point: 919° Density (g/mL): 2.3 Hygroscopic


  • Hygroscopic
  • -200 Mesh Powder

Formula: K₂MoO₄
MW: 238.13 g/mol
Melting Pt: 919 °C
Density: 2.3 g/mL
MDL Number: MFCD00011370
CAS Number: 13446-49-6
EINECS: 236-599-2

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR