N-(4-Bromobutyl)phthalimide ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-(4-Bromobutyl)isoindoline-1,3-dione

217125-100G 217125-25G 217125-5G
BT217125-100GEA 694 USD
BT217125-100G BT217125-25G BT217125-5G
N-(4-Bromobutyl)phthalimide ≥98%
N-(4-Bromobutyl)phthalimide

CAS: 5394-18-3; EC No: 226-401-2; MDL No: MFCD00005905 Powder; Molecular Formula: C12H12BrNO2; MW: 282.14 Melting Point: 76-80°; Boiling Point: 165-170°/1 mmHg


  • Powder

Formula: C₁₂H₁₂BrNO₂
MW: 282.14 g/mol
Boiling Pt: 165…170 °C/1 mmHg
Melting Pt: 76…80 °C
MDL Number: MFCD00005905
CAS Number: 5394-18-3
EINECS: 226-401-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR