Neocuproine hemihydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,9-Dimethyl-1,10-phenanthroline hemihydrate

217120-100G 217120-25G 217120-5G
BT217120-100GEA 847 USD
BT217120-100G BT217120-25G BT217120-5G
Neocuproine hemihydrate ≥98%
Neocuproine hemihydrate

CAS: 34302-69-7; EC No: 207-601-9; MDL No: MFCD00149306 Solid; Molecular Formula: C14H12N2·0.5H2O; MW: 217.27 Melting Point: 159-164°


  • Solid

Formula: C₂₈H₂₆N₄O
MW: 434.53 g/mol
Melting Pt: 159…164 °C
MDL Number: MFCD00149306
CAS Number: 34302-69-7
EINECS: 207-601-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR