Aluminium oxide, α-phase ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

217090-100G 217090-2KG 217090-500G
BT217090-100GEA 26 USD
BT217090-100G BT217090-2KG BT217090-500G
Aluminium oxide, α-phase ≥99.9% (trace metals basis)
Aluminium oxide, α-phase

CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424 20-50 Micron APS, Spherical Powder, S.A. 5-6 m2/g ; Molecular Formula: Al2O3; MW: 101.96 Melting Point: 2040°


  • 20-50 Micron APS, Spherical Powder, S.A. 5-6 m²/g

Formula: Al₂O₃
Melting Pt: 2040 °C
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR