Bromocresol purple sodium salt
Supplier: BeanTown Chemical
Synonyms:
Sodium α-(3-bromo-5-methyl-4-oxo-2,5-cyclohexadienylidene)-α-(3-bromo-5-methyl-4-hydroxyphenyl)toluenesulphonate, Bromocresol purple water soluble, Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-methylphenolate
217070-10G
217070-50G
BT217070-10GEA
39
USD
BT217070-10G
BT217070-50G
Bromocresol purple sodium salt
Bromocresol purple sodium salt
CAS: 62625-30-3; EC No: 263-655-3; MDL No: MFCD00148896
Powder; Molecular Formula: C21H15Br2NaO5S; MW: 562.20
Melting Point: 255° (decomposes)
- Powder
Formula:
C₂₁H₁₅Br₂NaO₅S Melting Pt: 255 °C (dec.) |
MDL Number:
MFCD00148896 CAS Number: 62625-30-3 EINECS: 263-655-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...