Methyl stearate ≥98%

Supplier: BeanTown Chemical

Synonyms: Stearic acid methyl ester, Methyl octadecanoate

217060-25G 217060-5G
BT217060-25GEA 182.57 USD
BT217060-25G BT217060-5G
Methyl stearate ≥98%
Methyl stearate

CAS: 112-61-8; EC No: 203-990-4; MDL No: MFCD00009005; RTECS: WI4460000 Solid; Linear Formula: CH3(CH2)16COOCH3; Molecular Formula: C19H38O2; MW: 298.51 Melting Point: 37-41°; Boiling Point: 181-182°/4 mmHg; Flash point: 110°C (230°F)


  • RTECS: WI4460000
  • Solid

Formula: CH₃(CH₂)₁₆CO₂CH₃
MW: 298.51 g/mol
Boiling Pt: 355
Melting Pt: 37…41 °C
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00009005
CAS Number: 112-61-8
EINECS: 203-990-4
Merck Index: 12,08959

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR