Carbonylhydridotris(triphenylphosphine)rhodium(I)

Supplier: BeanTown Chemical

Synonyms: Tris(triphenylphosphine)rhodium(I) Carbonyl Hydride, Carbonyltris(triphenylphosphine)rhodium(I) Hydride, Hydridocarbonyltris(triphenylphosphine)rhodium(I)

217015-1G 217015-250MG 217015-5G
BT217015-1GEA 251 USD
BT217015-1G BT217015-250MG BT217015-5G
Carbonylhydridotris(triphenylphosphine)rhodium(I)
Carbonylhydridotris(triphenylphosphine)rhodium(I)

CAS: 17185-29-4; EC No: 241-230-3; MDL No: MFCD00151644 Powder; Linear Formula: Rh(CO)H[P(C6H5)3]3; Molecular Formula: C55H46OP3Rh; MW: 918.78 Melting Point: 150° (decomposes) Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: C₅₅H₄₆OP₃Rh
MW: 918.80 g/mol
Melting Pt: 150 °C (dec.)
MDL Number: MFCD00151644
CAS Number: 17185-29-4
EINECS: 241-230-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR