Samarium(III) oxide ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
216985-10G
216985-50G
BT216985-10GEA
69
USD
BT216985-10G
BT216985-50G
Samarium(III) oxide ≥99.9% (trace metals basis)
Samarium(III) oxide
CAS: 12060-58-1; EC No: 235-043-6; MDL No: MFCD00011237
3-12mm Lump; Linear Formula: Sm2O3 ; MW: 348.72
Melting Point: 2325°
Density (g/mL): 8.35; Refractive Index: 1.97
- Refractive index: 1.97
- 3-12mm Lump
Formula:
O₃Sm₂ MW: 348.72 g/mol Melting Pt: 2325 °C Density: 8.35 g/mL |
MDL Number:
MFCD00011237 CAS Number: 12060-58-1 EINECS: 235-043-6 |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...