1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) ≥97%
Supplier: BeanTown Chemical
Synonyms:
3,4-Dihydronaphthalen-1(2H)-one
216980-100G
216980-500G
BT216980-100GEA
54
USD
BT216980-100G
BT216980-500G
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone) ≥97%
1,2,3,4-Tetrahydronaphthalen-1-one (α-tetralone)
CAS: 529-34-0; EC No: 208-460-6; MDL No: MFCD00001688; RTECS: QK4375000
Liquid; Molecular Formula: C10H10O; MW: 146.19
Melting Point: 2-7°; Boiling Point: 113-116°/6 mmHg; Flash point: 110°C (230°F)
Density (g/mL): 1.099; Refractive Index: 1.568
- RTECS: QK4375000
- Refractive index: 1.568
- Liquid
Formula:
C₁₀H₁₀O MW: 146.19 g/mol Boiling Pt: 113…116 °C/6 mmHg Melting Pt: 2…7 °C Density: 1.099 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00001688 CAS Number: 529-34-0 EINECS: 208-460-6 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...