1-n-Butyl-4-[(4-ethoxyphenyl)ethynyl]benzene ≥99%

Supplier: BeanTown Chemical

Synonyms: 1-Butyl-4-((4-ethoxyphenyl)ethynyl)benzene

216975-1G 216975-5G
BT216975-1GEA 61 USD
BT216975-1G BT216975-5G
1-n-Butyl-4-[(4-ethoxyphenyl)ethynyl]benzene ≥99%
1-n-Butyl-4-[(4-ethoxyphenyl)ethynyl]benzene

CAS: 85583-83-1; MDL No: MFCD01218016 Crystalline/Powder; Molecular Formula: C20H22O; MW: 278.39 Melting Point: 54°; Boiling Point: 168-170°/0.5 mmHg


  • Crystalline/Powder

Formula: C₂₀H₂₂O
MDL Number: MFCD01218016
CAS Number: 85583-83-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR