1,4-Diiodobenzene ≥98%

Supplier: BeanTown Chemical

216970-100G 216970-25G
BT216970-100GEA 172 USD
BT216970-100G BT216970-25G
1,4-Diiodobenzene ≥98%
1,4-Diiodobenzene

CAS: 624-38-4; EC No: 210-842-2; MDL No: MFCD00001054 Crystalline; Molecular Formula: C6H4I2; MW: 329.9 Melting Point: 131-133°; Boiling Point: 285° Light Sensitive


  • Light Sensitive
  • Crystalline

Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 285 °C
Melting Pt: 131…133 °C
MDL Number: MFCD00001054
CAS Number: 624-38-4
EINECS: 210-842-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR