(±)-Mercaptosuccinic acid ≥98%

Supplier: BeanTown Chemical

216965-100G 216965-25G 216965-500G
BT216965-100GEA 61.13 USD
BT216965-100G BT216965-25G BT216965-500G
(±)-Mercaptosuccinic acid ≥98%
(±)-Mercaptosuccinic acid

CAS: 70-49-5; EC No: 200-736-4; MDL No: MFCD00004860; RTECS: WM8225000 Solid; Linear Formula: HOOCCH(SH)CH2COOH; Molecular Formula: C4H6O4S; MW: 150.15 Melting Point: 155-157°


  • RTECS: WM8225000
  • Solid

Formula: C₄H₆O₄S
Melting Pt: 155…157 °C
MDL Number: MFCD00004860
CAS Number: 70-49-5
EINECS: 200-736-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR