Sodium benzenesulfinate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Benzenesulphinic acid sodium salt, Benzenesulphinic acid, sodium salt
216955-100G
216955-25G
216955-500G
BT216955-100GEA
63
USD
BT216955-100G
BT216955-25G
BT216955-500G
Sodium benzenesulfinate ≥98%
Sodium benzenesulphinate
CAS: 873-55-2; EC No: 212-842-8; MDL No: MFCD00013135; RTECS: DA9135000
Crystalline; Molecular Formula: C6H5NaO2S; MW: 164.16
Melting Point:
- RTECS: DA9135000
- Hygroscopic
- Crystalline
Formula:
C₆H₅NaO₂S MW: 164.16 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00013135 CAS Number: 873-55-2 EINECS: 212-842-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...