2-Bromo-3-hexylthiophene ≥98%
Supplier: BeanTown Chemical
216950-25G
216950-5G
BT216950-25GEA
536
USD
BT216950-25G
BT216950-5G
2-Bromo-3-hexylthiophene ≥98%
2-Bromo-3-hexylthiophene
CAS: 69249-61-2; MDL No: MFCD09907959
Liquid; Molecular Formula: C10H15BrS; MW: 247.20
Flash point: 110°C (230°F)
Density (g/mL): 1.24; Refractive Index: 1.529
- Refractive index: 1.529
- Liquid
Formula:
C₁₀H₁₅BrS Density: 1.24 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD09907959 CAS Number: 69249-61-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...