N,N-Diethylformamide ≥99%
Supplier: BeanTown Chemical
CAS: 617-84-5; EC No: 210-533-2; MDL No: MFCD00003287; RTECS: LQ1925000
Liquid; Linear Formula: HCON(CH2CH3)2; Molecular Formula: C5H11NO; MW: 101.15
Boiling Point: 176-177°; Flash point: 60°C (140°F)
Density (g/mL): 0.908; Refractive Index: 1.434
- RTECS: LQ1925000
- Refractive index: 1.434
- Liquid
Formula:
C₅H₁₁NO Boiling Pt: 176…177 °C Density: 0.908 g/mL Flash Pt: 60 °C (140 °F) |
MDL Number:
MFCD00003287 CAS Number: 617-84-5 EINECS: 210-533-2 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...