Terbium(III) acetylacetonate ≥99.9% (trace rare earth metals basis)

Supplier: BeanTown Chemical

216850-1G 216850-5G
BT216850-1GEA 35 USD
BT216850-1G BT216850-5G
Terbium(III) acetylacetonate ≥99.9% (trace rare earth metals basis)
Terbium(III) acetylacetonate

CAS: 14284-95-8; EC No: 238-195-1; MDL No: MFCD00013503 Powder; Molecular Formula: C15H24O6Tb ; MW: 459.27

Formula: C₁₅H₂₁O₆Tb
MDL Number: MFCD00013503
CAS Number: 14284-95-8

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR