5-Iodo-m-xylene ≥98%
Supplier: BeanTown Chemical
216845-10G
216845-50G
BT216845-10GEA
49
USD
BT216845-10G
BT216845-50G
5-Iodo-m-xylene ≥98%
5-Iodo-m-xylene
CAS: 22445-41-6; MDL No: MFCD00060659
Liquid; Molecular Formula: C8H9I; MW: 232.06
Boiling Point: 92-94°/3 mmHg; Flash point:
- Refractive index: 1.594
- Light Sensitive
- Liquid
Formula:
C₈H₉I Boiling Pt: 92…94 °C/3 mmHg Density: 1.608 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00060659 CAS Number: 22445-41-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...