Cesium wolframate ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Cesium tungsten oxide, Cesium tungstate

216765-25G 216765-5G
BT216765-25GEA 301.67 USD
BT216765-25G BT216765-5G
Cesium wolframate ≥99.9%
Cesium wolframate

CAS: 13587-19-4; EC No: 237-019-0; MDL No: MFCD00049438 -200 Mesh Powder; Linear Formula: Cs2WO4; MW: 513.66 Hygroscopic


  • Hygroscopic
  • -200 Mesh Powder

Formula: Cs₂O₄W
MW: 513.66 g/mol
MDL Number: MFCD00049438
CAS Number: 13587-19-4
EINECS: 237-019-0

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR