Disodium hydrogen citrate sesquihydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Citric acid disodium salt sesquihydrate, Sodium hydrogencitrate sesquihydrate, Disodium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate sesquihydrate, Sodium citrate dibasic sesquihydrate
216705-1KG
216705-250G
BT216705-1KGEA
139
USD
BT216705-1KG
BT216705-250G
Disodium hydrogen citrate sesquihydrate ≥99%
di-Sodium hydrogen citrate sesquihydrate
CAS: 6132-05-4; EC No: 205-623-3; MDL No: MFCD00150445
Crystalline; Molecular Formula: C6H6Na2O7·1.5H2O; MW: 263.11
Melting Point: 149° (decomposes)
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
C₆H₆Na₂O₇·1.5H₂O Melting Pt: 149 °C (dec.) |
MDL Number:
MFCD00150445 CAS Number: 6132-05-4 EINECS: 205-623-3 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...