4-Pyridone ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Hydroxypyridine, γ-Pyridone, 4-Pyridinol, Pyridin-4-ol, 1,4-Dihydro-4-Pyridinone

216625-100G 216625-25G 216625-500G
BT216625-100GEA 123.13 USD
BT216625-100G BT216625-25G BT216625-500G
4-Pyridone ≥97%
4-Pyridone

CAS: 626-64-2; EC No: 210-958-3; MDL No: MFCD00040458; RTECS: UU7701450 Powder; Molecular Formula: C5H5NO; MW: 95.10 Melting Point: 150-151°; Boiling Point: 230-235°/12 mmHg


  • RTECS: UU7701450
  • Powder

Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 230…235 °C/12 mmHg
Melting Pt: 150…151 °C
MDL Number: MFCD00006419
CAS Number: 626-64-2
EINECS: 210-958-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR