Silicon dioxide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

216605-100G 216605-25G
BT216605-100GEA 196 USD
BT216605-100G BT216605-25G
Silicon dioxide ≥99.9% (trace metals basis)
Silicon dioxide

CAS: 7631-86-9; EC No: 231-545-4; MDL No: MFCD00011232 0.5 Micron Powder; Linear Formula: SiO2; MW: 60.08 Melting Point: 1610°; Boiling Point: 2230° Density (g/mL): 2.2-2.6; Refractive Index: 1.544


  • Refractive index: 1.544
  • 0.5 Micron Powder

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: 1610 °C
Density: 2.2…2.6 g/mL
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR