Tris(3-methoxyphenyl)phosphine ≥98%
Supplier: BeanTown Chemical
216600-1G
216600-5G
BT216600-1GEA
80
USD
BT216600-1G
BT216600-5G
Tris(3-methoxyphenyl)phosphine ≥98%
Tris(3-methoxyphenyl)phosphine
CAS: 29949-84-6; MDL No: MFCD00008386
Solid; Linear Formula: (CH3OC6H4)3P; Molecular Formula: C21H21O3P; MW: 352.37
Melting Point: 113-115°
- Solid
Formula:
C₂₁H₂₁O₃P MW: 352.37 g/mol Melting Pt: 113…115 °C |
MDL Number:
MFCD00008386 CAS Number: 29949-84-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...