Yttrium(III) acetate tetrahydrate ≥99.9%

Supplier: BeanTown Chemical

216595-250G 216595-50G
BT216595-250GEA 178 USD
BT216595-250G BT216595-50G
Yttrium(III) acetate tetrahydrate ≥99.9%
Yttrium(III) acetate tetrahydrate

CAS: 85949-60-6; EC No: 245-612-0; MDL No: MFCD00013048 Powder; Molecular Formula: C6H9O6Y·4H2O; MW: 338.10 Density (g/mL): 1.5


  • Powder

Formula: C₆H₉O₆Y·4H₂O
MW: 338.10 g/mol (266.04 anhy)
Density: 1.5 g/mL
MDL Number: MFCD00013048
CAS Number: 85949-60-6
EINECS: 245-612-0

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR