Sodium acetate, anhydrous ≥99%
Supplier: BeanTown Chemical
Synonyms:
Acetic acid sodium salt
216570-1KG
216570-250G
216570-5KG
BT216570-1KGEA
63.43
USD
BT216570-1KG
BT216570-250G
BT216570-5KG
Sodium acetate, anhydrous ≥99%
Sodium acetate
CAS: 127-09-3; EC No: 204-823-8; MDL No: MFCD00012459; RTECS: AJ4300010
Solid; Linear Formula: CH3COONa; MW: 82.03
Melting Point: 300°; Flash point:
- RTECS: AJ4300010
- Hygroscopic
- Solid
Formula:
H₃CCOONa MW: 82.03 g/mol Boiling Pt: 881.4 Melting Pt: 324 Density: 1.528 g/mL Flash Pt: >250 °C (>482 °F) |
MDL Number:
MFCD00012459 CAS Number: 127-09-3 EINECS: 204-823-8 Merck Index: 12,08711 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...