Titanium(III) oxide ≥99.5%

Supplier: BeanTown Chemical

216545-250G 216545-50G
BT216545-250GEA 748 USD
BT216545-250G BT216545-50G
Titanium(III) oxide ≥99.5%
Titanium(III) oxide

CAS: 1344-54-3; EC No: 215-697-9; MDL No: MFCD00036278 -325 Mesh Powder; Linear Formula: Ti2O3; MW: 143.76 Melting Point: 2130° (decomposes) Density (g/mL): 4.49


  • -325 Mesh Powder

Formula: O₃Ti₂
MW: 143.76
Melting Pt: 1900 (dec.)
Density: 4.49 g/mL
MDL Number: MFCD00036278
CAS Number: 1344-54-3
EINECS: 215-697-9

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR