2,2,2-Trichloroethanol ≥99%

Supplier: BeanTown Chemical

Synonyms: 2,2,2-Trichloroethan-1-ol

216535-100G 216535-25G 216535-5G
BT216535-100GEA 50.01 USD
BT216535-100G BT216535-25G BT216535-5G
2,2,2-Trichloroethanol ≥99%
2,2,2-Trichloroethanol

CAS: 115-20-8; EC No: 204-071-0; MDL No: MFCD00004677; RTECS: KM3850000 Liquid; Linear Formula: Cl3CCH2OH ; MW: 149.40 Melting Point: 17.8°; Boiling Point: 152-154°; Flash point: 110°C (230°F) Density (g/mL): 1.56; Refractive Index: 1.49 Hygroscopic


  • RTECS: KM3850000
  • Refractive index: 1.49
  • Hygroscopic
  • Liquid

Formula: C₂H₃Cl₃O
MW: 149.4 g/mol
Boiling Pt: 152…154 °C
Melting Pt: 17.8 °C
Density: 1.56 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00004677
CAS Number: 115-20-8
EINECS: 204-071-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR