8-Amino-2-naphthol ≥98%

Supplier: BeanTown Chemical

216525-1G 216525-25G 216525-5G
BT216525-1GEA 26 USD
BT216525-1G BT216525-25G BT216525-5G
8-Amino-2-naphthol ≥98%
8-Amino-2-naphthol

CAS: 118-46-7; EC No: 204-252-4; MDL No: MFCD00004031; RTECS: QL3331000 Powder; Molecular Formula: C10H9NO; MW: 159.18 Melting Point: 206-210° Light Sensitive


  • RTECS: QL3331000
  • Light Sensitive
  • Powder

Formula: C₁₀H₉NO
Melting Pt: 206…210 °C
MDL Number: MFCD00004031
CAS Number: 118-46-7
EINECS: 204-252-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR