N,N-Bis(trifluoromethanesulfonyl)aniline ≥99%
Supplier: BeanTown Chemical
Synonyms:
1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N-Phenylbis(trifluoromethanesulphonimide)
216520-1G
216520-25G
216520-5G
BT216520-1GEA
24
USD
BT216520-1G
BT216520-25G
BT216520-5G
N,N-Bis(trifluoromethanesulfonyl)aniline ≥99%
N,N-Bis(trifluoromethanesulphonyl)aniline
CAS: 37595-74-7; MDL No: MFCD00000404
Crystalline/Powder; Linear Formula: C6H5N(SO2CF3)2; Molecular Formula: C8H5F6NO4S; MW: 357.25
Melting Point: 100-102°
Moisture Sensitive
- Moisture Sensitive
- Crystalline/Powder
Formula:
C₈H₅F₆NO₄S₂ MW: 357.25 g/mol Melting Pt: 100…102 °C |
MDL Number:
MFCD00000404 CAS Number: 37595-74-7 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...