4-Methyl-1-(2-nitrophenylsulfonyl)piperidine ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-[(4-methylpiperidyl)sulphonyl]-2-nitrobenzene, 4-Methyl-1-(2-nitro-benzenesulphonyl)-piperidine

216510-1G 216510-250MG
BT216510-1GEA 556 USD
BT216510-1G BT216510-250MG
4-Methyl-1-(2-nitrophenylsulfonyl)piperidine ≥97%
4-Methyl-1-(2-nitrophenylsulfonyl)piperidine

MDL No: MFCD00783740 Solid; Molecular Formula: C12H16N2O4S; MW: 284.33


  • Solid

Formula: C₁₂H₁₆N₂O₄S
MW: 284.33 g/mol
MDL Number: MFCD00783740
CAS Number: 346689-90-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR