Chromium(III) acetylacetonate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Tris(2,4-pentanedionato-O,O')chromium (III), Chromium (III) 2,4-pentanedionate, Chromium (III) tris(2,4-pentanedionato-O,O')
216495-100G
216495-500G
BT216495-100GEA
58.8
USD
BT216495-100G
BT216495-500G
Chromium(III) acetylacetonate ≥97%
Chromium(III) acetylacetonate
CAS: 21679-31-2; EC No: 244-526-0; MDL No: MFCD00000015; RTECS: GB7575000
Crystalline; Molecular Formula: C15H21CrO6; MW: 349.33
Melting Point: 210°; Boiling Point: 340°
- RTECS: GB7575000
- Crystalline
Formula:
C₁₅H₂₁CrO₆ MW: 349.32 g/mol Boiling Pt: 340 Melting Pt: 215.85 |
MDL Number:
MFCD00000015 CAS Number: 21679-31-2 EINECS: 244-526-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...