4-Bromo-N,N-dimethylaniline ≥98%

Supplier: BeanTown Chemical

216485-100G 216485-25G
BT216485-100GEA 112 USD
BT216485-100G BT216485-25G
4-Bromo-N,N-dimethylaniline ≥98%
4-Bromo-N,N-dimethylaniline

CAS: 586-77-6; EC No: 209-582-2; MDL No: MFCD00000093; RTECS: BW9300000 Crystalline; Molecular Formula: C8H10BrN; MW: 200.08 Melting Point: 52-54°; Boiling Point: 264°; Flash point: 110°C (230°F)


  • RTECS: BW9300000
  • Crystalline

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 264 °C
Melting Pt: 52…54 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00000093
CAS Number: 586-77-6
EINECS: 209-582-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR